Anti-HRH2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB39964-100EA 650 EUR
ABCAAB39964-100
Anti-HRH2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HRH2 (histamine receptor H2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR