Anti-MMP7 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB39984-100EA 672 EUR
ABCAAB39984-100
Anti-MMP7 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMP7 (matrix metallopeptidase 7 (matrilysin, uterine))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR