Anti-SDHB Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB39988-100EA 672 EUR
ABCAAB39988-100
Anti-SDHB Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SDHB (succinate dehydrogenase complex, subunit B, iron sulfur (Ip))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR