Anti-SCN5A Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB39992-100EA 666 EUR
ABCAAB39992-100
Anti-SCN5A Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SCN5A (sodium channel, voltage-gated, type V, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR