Anti-TGF beta induced factor 2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB39998-100EA 671 EUR
ABCAAB39998-100
Anti-TGF beta induced factor 2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TGF beta induced factor 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR