Anti-ADORA2B Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40002-100EA 688 EUR
ABCAAB40002-100
Anti-ADORA2B Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADORA2B (adenosine A2b receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR