Anti-ENPP1 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB40003-100EA 688 EUR
ABCAAB40003-100
Anti-ENPP1 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: ENPP1 (ectonucleotide pyrophosphatase/phosphodiesterase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR