Anti-TAZ Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40004-100EA 882 EUR
ABCAAB40004-100
Anti-TAZ Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAZ (tafazzin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR