Anti-FZD8 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB40012-100EA 688 EUR
ABCAAB40012-100
Anti-FZD8 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FZD8 (frizzled class receptor 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR