Anti-SLC2A1 Mouse monoclonal antibody [clone: SPM498]

Lieferant: Abcam
ABCAAB40084-500EA 768 EUR
ABCAAB40084-500
Anti-SLC2A1 Mouse monoclonal antibody [clone: SPM498]
Type: Primary
Antigen: SLC2A1 (solute carrier family 2 (facilitated glucose transporter), member 1)
Clonality: monoclonal
Clone: SPM498
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR