Anti-RUNX3 Mouse Monoclonal Antibody [clone: R3-5G4]

Lieferant: Abcam
ABCAAB40278-100EA 703 EUR
ABCAAB40278-100
Anti-RUNX3 Mouse Monoclonal Antibody [clone: R3-5G4]
Antikörper
Type: Primary
Antigen: RUNX3 (runt-related transcription factor 3)
Clonality: monoclonal
Clone: R3-5G4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR