Anti-Lymphocyte Activation Gene 3 Mouse Monoclonal Antibody [clone: 11E3]

Lieferant: Abcam
ABCAAB40465-50EA 921 EUR
ABCAAB40465-50
Anti-Lymphocyte Activation Gene 3 Mouse Monoclonal Antibody [clone: 11E3]
Antikörper
Type: Primary
Antigen: Lymphocyte Activation Gene 3
Clonality: monoclonal
Clone: 11000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR