Anti-DRD3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB40655-100EA 703 EUR
ABCAAB40655-100
Anti-DRD3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DRD3 (dopamine receptor D3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR