Anti-DRD5 Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB40656-100EA 590 EUR
ABCAAB40656-100
Anti-DRD5 Rabbit polyclonal antibody
Type: Primary
Antigen: DRD5 (dopamine receptor D5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR