Anti-IGF1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB40657-100EA 868 EUR
ABCAAB40657-100
Anti-IGF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IGF1 (insulin-like growth factor 1 (somatomedin C))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR