Anti-IL15 Rabbit Monoclonal Antibody [clone: EP433Y]

Lieferant: Abcam
ABCAAB40668-100EA 877 EUR
ABCAAB40668-100
Anti-IL15 Rabbit Monoclonal Antibody [clone: EP433Y]
Antikörper
Type: Primary
Antigen: IL15 (interleukin 15)
Clonality: monoclonal
Clone: EP433Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR