Anti-RHO Rabbit Monoclonal Antibody [clone: EP487Y]
Lieferant: Abcam
ABCAAB40673-100EA 903 EUR
ABCAAB40673-100
Anti-RHO Rabbit Monoclonal Antibody [clone: EP487Y]
Antikörper
Type: Primary
Antigen: RHO (rhodopsin)
Clonality: monoclonal
Clone: EP487Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen