Anti-TIA1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB40693-100EA 738 EUR
ABCAAB40693-100
Anti-TIA1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIA1 (TIA1 cytotoxic granule-associated RNA binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR