Anti-KLK2 Mouse Monoclonal Antibody [clone: 6B7]

Lieferant: Abcam
ABCAAB40749-100EA 682 EUR
ABCAAB40749-100
Anti-KLK2 Mouse Monoclonal Antibody [clone: 6B7]
Antikörper
Type: Primary
Antigen: KLK2 (kallikrein-related peptidase 2)
Clonality: monoclonal
Clone: 6B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR