Anti-BCL10 Rabbit monoclonal antibody unconjugated [clone: ep605y]

Lieferant: Abcam
ABCAAB40752-100EA 672 EUR
ABCAAB40752-100
Anti-BCL10 Rabbit monoclonal antibody unconjugated [clone: ep605y]
Antikörper
Type: Primary
Antigen: BCL10 (B-cell CLL/lymphoma 10)
Clonality: monoclonal
Clone: ep605y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR