Anti-MAP3K3 Rabbit Monoclonal Antibody [clone: ep600y]

Lieferant: Abcam
ABCAAB40756-100EA 697 EUR
ABCAAB40756-100
Anti-MAP3K3 Rabbit Monoclonal Antibody [clone: ep600y]
Antikörper
Type: Primary
Antigen: MAP3K3 (mitogen-activated protein kinase kinase kinase 3)
Clonality: monoclonal
Clone: ep600y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR