Anti-INA Rabbit Monoclonal Antibody [clone: EP676Y]
Lieferant: Abcam
ABCAAB40758-100EA 914 EUR
ABCAAB40758-100
Anti-INA Rabbit Monoclonal Antibody [clone: EP676Y]
Antikörper
Type: Primary
Antigen: INA (internexin neuronal intermediate filament protein, alpha)
Clonality: monoclonal
Clone: EP676Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen