Anti-alpha Adducin Rabbit Monoclonal Antibody [clone: EP734Y]

Lieferant: Abcam
ABCAAB40760-100EA 688 EUR
ABCAAB40760-100
Anti-alpha Adducin Rabbit Monoclonal Antibody [clone: EP734Y]
Antikörper
Type: Primary
Antigen: alpha Adducin
Clonality: monoclonal
Clone: EP734Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR