Anti-CEBPA Rabbit monoclonal antibody unconjugated [clone: EP708Y]

Lieferant: Abcam
ABCAAB40761-100EA 757 EUR
ABCAAB40761-100
Anti-CEBPA Rabbit monoclonal antibody unconjugated [clone: EP708Y]
Antikörper
Type: Primary
Antigen: CEBPA (CCAAT/enhancer binding protein (C/EBP), alpha)
Clonality: monoclonal
Clone: EP708Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR