Anti-EEF2 Rabbit monoclonal antibody unconjugated [clone: EP723Y]

Lieferant: Abcam
ABCAAB40762-100EA 672 EUR
ABCAAB40762-100
Anti-EEF2 Rabbit monoclonal antibody unconjugated [clone: EP723Y]
Antikörper
Type: Primary
Antigen: EEF2 (eukaryotic translation elongation factor 2)
Clonality: monoclonal
Clone: EP723Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR