Anti-PTPRC Rabbit Monoclonal Antibody [clone: EP322Y]

Lieferant: Abcam
ABCAAB40763-100EA 796 EUR
ABCAAB40763-100
Anti-PTPRC Rabbit Monoclonal Antibody [clone: EP322Y]
Antikörper
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: EP322Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR