Anti-JUN Rabbit Monoclonal Antibody [clone: EP693Y]

Lieferant: Abcam
ABCAAB40766-100EA 688 EUR
ABCAAB40766-100
Anti-JUN Rabbit Monoclonal Antibody [clone: EP693Y]
Antikörper
Type: Primary
Antigen: JUN (jun proto-oncogene)
Clonality: monoclonal
Clone: EP693Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR