Anti-PARD3 Rabbit Monoclonal Antibody [clone: EP310Y]
Lieferant: Abcam
ABCAAB40769-100EA 936 EUR
ABCAAB40769-100
Anti-PARD3 Rabbit Monoclonal Antibody [clone: EP310Y]
Antikörper
Type: Primary
Antigen: PARD3 (par-3 family cell polarity regulator)
Clonality: monoclonal
Clone: EP310Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen