Anti-SMAD5 Rabbit Monoclonal Antibody [clone: EP619Y]

Lieferant: Abcam
ABCAAB40771-100EA 713 EUR
ABCAAB40771-100
Anti-SMAD5 Rabbit Monoclonal Antibody [clone: EP619Y]
Antikörper
Type: Primary
Antigen: SMAD5 (SMAD family member 5)
Clonality: monoclonal
Clone: EP619Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR