Anti-CDH1 Rabbit Monoclonal Antibody [clone: EP700Y]

Lieferant: Abcam
ABCAAB40772-100EA 793 EUR
ABCAAB40772-100
Anti-CDH1 Rabbit Monoclonal Antibody [clone: EP700Y]
Antikörper
Type: Primary
Antigen: CDH1 (cadherin 1, type 1, E-cadherin (epithelial))
Clonality: monoclonal
Clone: EP700Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR