Anti-CDK5 Rabbit monoclonal antibody unconjugated [clone: EP715Y]

Lieferant: Abcam
ABCAAB40773-100EA 999 EUR
ABCAAB40773-100
Anti-CDK5 Rabbit monoclonal antibody unconjugated [clone: EP715Y]
Antikörper
Type: Primary
Antigen: CDK5 (cyclin-dependent kinase 5)
Clonality: monoclonal
Clone: EP715Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR