Anti-PI 3 Kinase catalytic subunit alpha Rabbit Monoclonal Antibody [clone: EP383Y]

Lieferant: Abcam
ABCAAB40776-100EA 757 EUR
ABCAAB40776-100
Anti-PI 3 Kinase catalytic subunit alpha Rabbit Monoclonal Antibody [clone: EP383Y]
Antikörper
Type: Primary
Antigen: PI 3 Kinase catalytic subunit alpha
Clonality: monoclonal
Clone: EP383Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR