Anti-IRS1 Rabbit Monoclonal Antibody [clone: EP263Y]

Lieferant: Abcam
ABCAAB40777-100EA 697 EUR
ABCAAB40777-100
Anti-IRS1 Rabbit Monoclonal Antibody [clone: EP263Y]
Antikörper
Type: Primary
Antigen: IRS1 (insulin receptor substrate 1)
Clonality: monoclonal
Clone: EP263Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR