Anti-BCR Rabbit monoclonal antibody unconjugated [clone: EP535Y]

Lieferant: Abcam
ABCAAB40779-100EA 688 EUR
ABCAAB40779-100
Anti-BCR Rabbit monoclonal antibody unconjugated [clone: EP535Y]
Antikörper
Type: Primary
Antigen: BCR (breakpoint cluster region)
Clonality: monoclonal
Clone: EP535Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR