Anti-SYK Rabbit Monoclonal Antibody [clone: EP573Y]

Lieferant: Abcam
ABCAAB40781-100EA 757 EUR
ABCAAB40781-100
Anti-SYK Rabbit Monoclonal Antibody [clone: EP573Y]
Antikörper
Type: Primary
Antigen: SYK (spleen tyrosine kinase)
Clonality: monoclonal
Clone: EP573Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR