Anti-Insulin Receptor R Rabbit Monoclonal Antibody [clone: EP327Y]

Lieferant: Abcam
ABCAAB40782-100EA 857 EUR
ABCAAB40782-100
Anti-Insulin Receptor R Rabbit Monoclonal Antibody [clone: EP327Y]
Antikörper
Type: Primary
Antigen: Insulin Receptor R
Clonality: monoclonal
Clone: EP327Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR