Anti-MAP2K4 Rabbit Monoclonal Antibody [clone: EP551Y]

Lieferant: Abcam
ABCAAB40784-100EA 672 EUR
ABCAAB40784-100
Anti-MAP2K4 Rabbit Monoclonal Antibody [clone: EP551Y]
Antikörper
Type: Primary
Antigen: MAP2K4 (mitogen-activated protein kinase kinase 4)
Clonality: monoclonal
Clone: EP551Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR