Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 209F2]

Lieferant: Abcam
ABCAAB40787-500EA 634 EUR
ABCAAB40787-500
Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 209F2]
Antikörper
Type: Primary
Antigen: Clostridium botulinum E Toxoid
Clonality: monoclonal
Clone: 209F2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR