Anti-PTK2 Rabbit Monoclonal Antibody [clone: EP695Y]
Lieferant: Abcam
ABCAAB40794-100EA 993 EUR
ABCAAB40794-100
Anti-PTK2 Rabbit Monoclonal Antibody [clone: EP695Y]
Antikörper
Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: monoclonal
Clone: EP695Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen