Anti-PAK1 Rabbit Monoclonal Antibody [clone: EP656Y]

Lieferant: Abcam
ABCAAB40795-100EA 727 EUR
ABCAAB40795-100
Anti-PAK1 Rabbit Monoclonal Antibody [clone: EP656Y]
Antikörper
Type: Primary
Antigen: PAK1 (p21 protein (Cdc42/Rac)-activated kinase 1)
Clonality: monoclonal
Clone: EP656Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR