Anti-DARPP32 Rabbit monoclonal antibody unconjugated [clone: EP720Y]

Lieferant: Abcam
ABCAAB40801-100EA 763 EUR
ABCAAB40801-100
Anti-DARPP32 Rabbit monoclonal antibody unconjugated [clone: EP720Y]
Antikörper
Type: Primary
Antigen: DARPP32
Clonality: monoclonal
Clone: EP720Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR