Anti-CDKN2A Rabbit monoclonal antibody unconjugated [clone: EP435Y]

Lieferant: Abcam
ABCAAB40803-100EA 688 EUR
ABCAAB40803-100
Anti-CDKN2A Rabbit monoclonal antibody unconjugated [clone: EP435Y]
Antikörper
Type: Primary
Antigen: CDKN2A (cyclin-dependent kinase inhibitor 2A)
Clonality: monoclonal
Clone: EP435Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR