Anti-CD3 zeta Rabbit Monoclonal Antibody [clone: EP286Y]

Lieferant: Abcam
ABCAAB40804-100EA 672 EUR
ABCAAB40804-100
Anti-CD3 zeta Rabbit Monoclonal Antibody [clone: EP286Y]
Antikörper
Type: Primary
Antigen: CD3 zeta
Clonality: monoclonal
Clone: EP286Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR