Anti-MAPK7 Rabbit Monoclonal Antibody [clone: EP791Y]

Lieferant: Abcam
ABCAAB40809-100EA 688 EUR
ABCAAB40809-100
Anti-MAPK7 Rabbit Monoclonal Antibody [clone: EP791Y]
Antikörper
Type: Primary
Antigen: MAPK7 (mitogen-activated protein kinase 7)
Clonality: monoclonal
Clone: EP791Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR