Anti-Natriuretic Peptide Receptor A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB40817-100EA 672 EUR
ABCAAB40817-100
Anti-Natriuretic Peptide Receptor A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Natriuretic Peptide Receptor A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR