Anti-DARC Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB40821-100EA 672 EUR
ABCAAB40821-100
Anti-DARC Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR