Anti-CBX1 / HP1 beta Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40828-100EA 672 EUR
ABCAAB40828-100
Anti-CBX1 / HP1 beta Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CBX1 / HP1 beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR