Anti-EPB41L3 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB40831-100EA 666 EUR
ABCAAB40831-100
Anti-EPB41L3 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EPB41L3 (erythrocyte membrane protein band 4.1-like 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR