Anti-RPS19 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40833-100EA 672 EUR
ABCAAB40833-100
Anti-RPS19 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPS19 (ribosomal protein S19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR