Anti-ADRB2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40834-100EA 672 EUR
ABCAAB40834-100
Anti-ADRB2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADRB2 (adrenoceptor beta 2, surface)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR